Learn More
Fisher Bioreagents Sucrose gel loading dye for DNA gels, 5X, contains 40% sucrose, DNase and RNase free for electrophoresis
Brand:  Fisher Bioreagents BP655-1
Sucrose gel loading dye for DNA gels, 5X, contains 40% sucrose, DNase and RNase free for electrophoresis
Code : 32
Additional Details : Weight : 0.05100kg
Product Code. 10306743