missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Sucrose, Extra Pure, SLR
Specifications
Melting Point | 190°C |
Color | White |
pH | 6.5 to 7.5 |
Packaging | Plastic powder jar |
Quantity | 3 kg |
Formula Weight | 342.29g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'