missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Catalog Number 10346150
33.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


19.75 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


33.40 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

C12H22O11, CAS Number-57-50-1, amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar, 1kg, CHEBI:17992, White, 342.29g/mol, CZMRCDWAGMRECN-UGDNZRGBSA-N

TRUSTED_SUSTAINABILITY
Melting Point 190°C
Color White
pH 6.5 to 7.5
Packaging Plastic powder jar
Quantity 500 g
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Formula Weight 342.29g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'