missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
115.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
C12H22O11, CAS Number-57-50-1, amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar, 1kg, CHEBI:17992, White, 342.29g/mol, CZMRCDWAGMRECN-UGDNZRGBSA-N
Specifications
Melting Point | 190°C |
Color | White |
pH | 6.5 to 7.5 |
Packaging | Plastic powder jar |
Quantity | 3 kg |
IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Formula Weight | 342.29g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'