missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, 99.97%, MP Biomedicals™
Click to view available options
Quantity:
5 kg
Specifications
Color | White |
Density | 1.587g/cm3 at 25°C |
Assay Percent Range | 99.7% |
Quantity | 5 kg |
IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Formula Weight | 342.3 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Sucrose |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'