missing translation for 'onlineSavingsMsg'
Learn More

Suberic acid bis(N-hydroxysuccinimide ester), 97%, Thermo Scientific Chemicals

Catalog Number 10524944
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
68528-80-3
C16H20N2O8
368.34
MFCD00049059
ZWIBGKZDAWNIFC-UHFFFAOYSA-N
disuccinimidyl suberate, bis 2,5-dioxopyrrolidin-1-yl octanedioate, suberic acid bis n-hydroxysuccinimide ester, di n-succinimidyl suberate, bicl100, 2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, nhs-sa, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester
100658
bis(2,5-dioxopyrrolidin-1-yl) octanedioate
C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O
This item is not returnable. View return policy
68528-80-3
C16H20N2O8
368.34
MFCD00049059
ZWIBGKZDAWNIFC-UHFFFAOYSA-N
disuccinimidyl suberate, bis 2,5-dioxopyrrolidin-1-yl octanedioate, suberic acid bis n-hydroxysuccinimide ester, di n-succinimidyl suberate, bicl100, 2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, nhs-sa, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester
100658
bis(2,5-dioxopyrrolidin-1-yl) octanedioate
C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 368.34
Percent Purity 97%
Chemical Name or Material Suberic acid bis(N-hydroxysuccinimide ester)
compliance-icons
Product Identifier
  • Suberic acid bis(N-hydroxysuccinimide ester)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'