missing translation for 'onlineSavingsMsg'
Learn More

SU 5416, Tocris Bioscience™

VEGFR inhibitor. Also inhibits KIT, RET, MET and FLT3

Brand:  Tocris (Bio-Techne) 303710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452274

  • £157.29 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of vascular endothelial growth factor receptor (VEGFR) that also inhibits other tyrosine kinases KIT, MET, FLT3 and RET. Displays no activity against EGFR, HER2, IGF1R and PDGFR. Inhibits tumor vascularization and growth of multiple tumor types.
Specifications

Chemical Identifiers

204005-46-9
238.29
semaxanib, semaxinib, semoxind, semaxanib su5416, semaxnib, tsu 16, unii-71ia9s35aj, vegf receptor 2 kinase inhibitor iii, tsu-16, semaxanib usan/inn
(3Z)-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylidene]-1H-indol-2-one
C15H14N2O
WUWDLXZGHZSWQZ-WQLSENKSSA-N
5329098
CC1=CC(=C(N1)C=C2C3=CC=CC=C3NC2=O)C

Specifications

10mg
204005-46-9
semaxanib, semaxinib, semoxind, semaxanib su5416, semaxnib, tsu 16, unii-71ia9s35aj, vegf receptor 2 kinase inhibitor iii, tsu-16, semaxanib usan/inn
CC1=CC(=C(N1)C=C2C3=CC=CC=C3NC2=O)C
238.29
238.28
SU 5416
C15H14N2O
WUWDLXZGHZSWQZ-WQLSENKSSA-N
(3Z)-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylidene]-1H-indol-2-one
5329098
>99%
Videos
Documents