Learn More
Tocris Bioscience™ SU 3327
Selective JNK inhibitor
Brand:  Tocris Bioscience™ 360750
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433284
  • £699.72 / 50mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Selective inhibitor of c-Jun N-terminal kinase (JNK) (IC50 = 0.7 μM). Inhibits the protein-protein interaction between JNK and JIP (IC50 = 239 nM). Displays selectivity over p38 MAPK and Akt. Restores insulin sensitivity in a mouse model of type-2 diabetes.
Specifications
Specifications
SU 3327
>99
50mg
SU 3327
261.3