missing translation for 'onlineSavingsMsg'
Learn More

Styrene-d{8}, 98% (Isotopic) stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog Number p-7058956
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
19361-62-7
C8H8
112.201
MFCD00044231
PPBRXRYQALVLMV-GDALLCCDSA-N
styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
88025
1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
C=CC1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
19361-62-7
C8H8
112.201
MFCD00044231
PPBRXRYQALVLMV-GDALLCCDSA-N
styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
88025
1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
C=CC1=CC=CC=C1

Styrene-d{8} is used in Organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.979 g/mL
Flash Point 31°C (88°F)
Packaging Ampule
Refractive Index 1.5445
Sensitivity Air
Quantity 1 g
Boiling Point 50°C to 51°C (25mmHg)
Linear Formula C6D5CD=CD2
UN Number UN2055
Beilstein 2327045
Solubility Information It is soluble in water 0.24g/L (20°C).
Formula Weight 112.2
Percent Purity 98%
Chemical Name or Material Styrene-d8, Stabilized with 4-tert-butylcatechol, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'