Learn More
Cole-Parmer™ V Series Stuart™ Vortex Mixer
This V Series Stuart™ Vortex Mixer is a compact unit perfect for individual work stations.
Brand: Cole-Parmer™ 04729-01
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Mix a wide variety of tube sizes and shapes with the included rubber cup head.
- Heavy die-cast body won't dance across your bench.
- Automatic start when the soft rubber cup is pressed down.
- The Vortex mixer provides rapid mixing of samples contained in test-tubes, small flasks and bottles.
- Starts automatically when the rubber cup is depressed and stops once vessel is removed.
- Control the mixing speed by selecting from 200 rpm for more gentle mixing to 2500 rpm for vigorous agitation.
- Heavy die-cast body for stability during use.
- Includes integral retort rod for securing vessels above the vortex action for long-term mixing.
Specifications
Fixed Speed | |
Vortex Mixer |