Learn More
Bibby Scientific™ Stuart™ Hotplate
Heats uniformly and accommodates multiple vessels or microscope slides with large capacity. Bibby Scientific™ Stuart™ Hotplates with digital setting and microprocessor control provide accurate monitoring of the plate temperature.
£716.21 - £866.93
Specifications
Max. Temperature (Metric) 300°C
Voltage 230V
Hertz 50-60Hz
Control Digital
Controller Microprocessor
View More Specs
Products 2
Product Code Brand Height (Metric) Width (Metric) Depth (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Width (Metric) Depth (Metric) Price Quantity & Availability  
11426518 Bibby Scientific™
SD500
130mm 520mm 360mm £866.93
Each
In Stock 11
Add to basket
View alternative products »
12048323 Bibby Scientific™
SD300
105mm 320mm 365mm £716.21
Each
In Stock 3
Add to basket
Description
Description
  • Digital setting and control of plate temperature
  • Accurate temperature control by microprocessor
  • Easy to use controls
  • Metal top plate gives very even plate temperature and uniform heating conditions.
  • Hotplate with microprocessor control for accurate monitoring of the plate temperature.
  • Large capacity to accommodate multiple vessels or microscope slides.
Specifications
Specifications
300°C
50-60Hz
Microprocessor
230V
Digital