Learn More
Bibby Scientific™ Stuart™ Glass Ceramic Hotplate
Features a glass ceramic top with excellent chemical resistance. Bibby Scientific™ Stuart™ Glass Ceramic Hotplates have an easy to clean surface clean with high plate temperature settings for faster heating.
£287.55 - £2320.00
Specifications
Top Plate Material Glass Ceramic
Voltage 230V
Hertz 50-60Hz
View More Specs
Products 4
Product Code Brand Max. Temperature (Metric) Height (Metric) Width (Metric) Electrical Requirements Price Quantity & Availability  
Product Code Brand Max. Temperature (Metric) Height (Metric) Width (Metric) Electrical Requirements Price Quantity & Availability  
11436518 Bibby Scientific™
CB500
375°C 130mm 520mm 230V, 50-60Hz, 1200W £745.07
Each View alternative products
Save Up to 
»
In Stock 2
Add to basket
11476508 Bibby Scientific™
CB300
450°C 105mm 300mm 230V, 50-60Hz, 2250W £595.00
Each View alternative products
Save Up to 
»
Estimated Shipment
03-04-2019
Add to basket
12865916 View accessory products Bibby Scientific™
UC150
450°C 122mm 172mm 230V, 50-60Hz, 500W £287.55
Each View alternative products
Save Up to 
»
Estimated Shipment
01-04-2019
Add to basket
11456638 Bibby Scientific™
CP300
400°C 65mm 150mm 230V, 50-60Hz, 900W £2320.00
Each View alternative products
Save Up to 
»
Estimated Shipment
03-04-2019
Add to basket
Description
Description
Features a glass ceramic top with excellent chemical resistance. The surface is easy to clean with high plate temperature settings for faster heating. Possesses an integral fitting for a retort rod and the bottom is shaped to allow a retort base to slide underneath the unit if required so saving further bench space.
  • Chemically resistant ceramic top plate
  • Flashing 'Hot' warning light, mains independent
  • Accurate temperature control with LED setting scale
  • Compact space saving design
  • IP rating 32
Specifications
Specifications
Glass Ceramic
230V
50-60Hz