Learn More
Techne™ Stuart™ Borosilicate Glass Culture Flask
Culture flask for magnetic stirrers (without end caps, seals or stirrer rods)
£195.00
Specifications
For Use With (Equipment) Biological Stirrers Glass no End Caps, Seals or Stirrer Rods
Material Borosilicate Glass
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12927286
View Documents Promotion Details
Techne™
F7987
125 mL
£195.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-10-2022
to see stock.
Add to basket
12937286
View Documents Promotion Details
Techne™
F7690
250 mL
£195.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-10-2022
to see stock.
Add to basket
Description
Description
The gentle stirring action aids attachment and promotes cell growth. Soft-start electronic acceleration control avoids turbulence and thus minimises cell damage. Interval stirring with variable 'ON-OFF' times improves cell attachment rate, While continuous stirring is controllable up to 80rpm. All units may be used in an incubator or warm room. Culture vessels are made from siliconised borosilicate glass, to minimise cell attachment to the vessels. Electrical supply 230V, 50Hz, 2W. Designed for the cultivation of cells on microcarrier beads. This system can also be used for suspension cell culture techniques.
Specifications
Specifications
Biological Stirrers Glass no End Caps, Seals or Stirrer Rods
Borosilicate Glass