Learn More
Cole-Parmer™ Essentials Replacement Cell Culturing Flask
Enhance the life of your BS-200 series cell culture stirring systems with this Replacement Cell Culturing Flask.
Brand: Cole-Parmer™ Essentials 21837-35
Code : 04
Additional Details : Weight : 5.00000kg
Description
- Constructed of borosilicate glass
The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Replacement flasks do not include caps or stirring mechanisms.
Specifications
250 mL | |
Borosilicate Glass | |
Biological Stirrers Glass no End Caps, Seals or Stirrer Rods | |
Flat |