missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Strontium titanium oxide, 40% in H2O colloidal dispersion

Brand:  Alfa Aesar™ 045845.22

Code : NEW

Additional Details : CAS Number : 12060-59-2 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15407105

  • £111.41 / 100g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Strontium titanium oxide
Colloidal dispersion
SrTiO3
MFCD00049554
12060-59-2
O3SrTi
183.51

Specifications

Strontium titanium oxide
12060-59-2
O3SrTi
183.51
40% in H2O
100g
Colloidal dispersion
Dry Powder
SrTiO3
MFCD00049554
0.05 to 0.15 Micron APS
Videos
SDS