missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number 11318827
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7759-02-6
CHEBI:63041
This item is not returnable. View return policy
7759-02-6
CHEBI:63041

Strontium sulfate is used as a precursor to other strontium compounds and as ceramic precursor. Its application in nitrate for use in pyrotechnics. Used as a fining agent in crystal glass. It is also used in powder coating, liquid paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1600°C
Physical Form Crystalline
Quantity 25 g
Molecular Formula Cl2Mn
MDL Number MFCD00011114
Merck Index 14,8848
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
Solubility Information Soluble in dilute hydrochloric acid and nitric acid . Slightly soluble in water < 0.01g/L . Insoluble in ethanol and alkalis.
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Mn++]
IUPAC Name manganese(2+) dichloride
Molecular Weight (g/mol) 125.84
PubChem CID 10313134
Formula Weight 183.68
Grade Reagent
Density 3.960 g/mL
Chemical Name or Material Strontium sulfate, For reagent grade
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'