missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium iodide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
58.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Strontium iodide is used as a scintillation gamma radiation detector. It is also used in a range of hand-held radiation detection instruments as well as in medical, industrial and environmental applications. It is used in medicine as a substitute for potassium iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 515°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 5 g |
Merck Index | 14,8844 |
Solubility Information | Soluble in water and ethanol. |
Formula Weight | 341.43 |
Percent Purity | 99.99% |
Odor | Odorless |
Sensitivity | Hygroscopic; Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'