missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Chloride, 6-Hydrate, Low in Magnesium, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
401.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Physical Form | Crystals |
Quantity | 500 g |
Molecular Formula | SrCl2·6H2O |
Formula Weight | 266.62 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.96kg |
Chemical Name or Material | Strontium Chloride, 6-Hydrate, Low in Magnesium |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'