missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium acetate hemihydrate, Reagent Grade, Thermo Scientific Chemicals
34.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Strontium acetate is used as a catalyst, as a chemical intermediate and in medicine. The primary application of strontium acetate is as an active component in toothpastes. This compound is used to desensitize teeth to extremes of hot and cold, sweets, acids, or touch.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 150°C (in -0.5H2O) |
Physical Form | Crystalline |
Quantity | 250 g |
Merck Index | 14,8836 |
Solubility Information | Soluble in water and slightly soluble in alcohol. |
Formula Weight | 214.72 (205.71 Anhydrous) |
Grade | Reagent |
Density | 2.099 |
Chemical Name or Material | Strontium acetate hemihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'