missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Strontium 86, 86Sr, plasma standard solution, Specpure™, 86Sr 10μg/mL

Brand:  Alfa Aesar™ 045286.AD

Code : NEW

Additional Details : Weight : 0.00050kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 50mL

Product Code. 15486875

  • £117.20 / 50mL


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Specifications

Strontium 86, 86Sr, plasma standard solution
Specpure™
MFCD01075356
50mL
86Sr 10μg/mL
Liquid
UN3264
Matrix: 2% HNO3
Videos
SDS