missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550255
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total stress-associated endoplasmic reticulum protein 1 protein.
SERP1 Polyclonal antibody specifically detects SERP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SERP1 | |
Polyclonal | |
Unconjugated | |
SERP1 | |
D3Ucla1; RAMP4; ribosome associated membrane protein 4; ribosome-attached membrane protein 4; Serp1; Stress-associated endoplasmic reticulum protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27230, 28146, 80881 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9R2C1, Q9Y6X1, Q9Z1W5 | |
SERP1 | |
Fusion protein corresponding to residues near the N terminal of human stress-associated endoplasmic reticulum protein 1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |