Learn More
Invitrogen™ eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 46-4317-82
Code : Z2
Additional Details : Weight : 0.25000kg
Specifications
0.2 mg/mL | |
100 μg | |
eBioscience™ | |
Biotin |
PerCP-eFluor 710 | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
PerCP-eFluor 710 |
For Research Use Only