missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PerCP-eFluor™ 710

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Brand:  Invitrogen™ 46-4317-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15301320

  • £85.18 / 25µg


EU Stock 2
Add to basket

Specifications

Specifications

0.2mg/mL
Flow Cytometric Analysis
2° to 8°C. Do not freeze. Light-sensitive material.
SA, Sav
PerCP-eFluor 710
25μg
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Videos

For Research Use Only