missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PerCP-Cyanine5.5

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Brand:  Invitrogen™ 45-4317-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15321180

  • £157.88 / 100µg


EU Stock 6
Add to basket

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Specifications

Specifications

0.2mg/mL
Flow Cytometric Analysis
2° to 8°C. Do not freeze. Light sensitive material.
SA, Sav
PerCP-Cyanine5.5
100μg
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Videos