Learn More
Invitrogen™ eBioscience™ Streptavidin PE-eFluor™ 610 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 61-4317-82
Code : NEW
Additional Details : Weight : 0.09500kg
Alert:
Contains Sodium azide
Specifications
0.2 mg/mL | |
100 μg | |
eBioscience™ | |
Biotin |
PE-eFluor 610 | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
PE-eFluor 610 |