missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PE-eFluor™ 610

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Brand:  Invitrogen™ 61-4317-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15530497

  • £62.64 / 25µg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Alert:

Contains Sodium azide


Specifications

Specifications

0.2mg/mL
Flow Cytometric Analysis
2° to 8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling & experimental procedures.
SA, Sav
PE-eFluor 610
25μg
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Videos