Learn More
Molecular Probes™ Streptavidin HRP (ELISA GD)
Streptavidin HRP (ELISA GD)
Brand: Molecular Probes™ SNN2004
Code : 50
Additional Details : Weight : 0.05250kg
Description
Purified from Streptomyces avidinii and provided in a phosphate buffered saline solution. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.Format/Formulation: HRP Conjugate (ELISA grade)
Specifications
1 mg/mL | |
Store in freezer (-5 to -30°C). | |
Liquid | |
HRP (Horseradish Peroxidase) |
HRP | |
Approved for shipment on Wet or Dry Ice | |
Streptavidin Conjugate (fluorescent) |
For Laboratory Use