missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Streptavidin-HRP

Product Code. 17136781
Click to view available options
Quantity:
1 mg
Unit Size:
1mg

Product Code. 17136781

Brand: ACROBiosystems STNNH1151mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Streptavidin is a tetrameric protein purified from the bacterium Streptomyces avidinii, and exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit. Streptavidin (PI=6.0-7.5) has lower level of non-specific binding to various biological components at physiological pH than avidin (PI=7.4), resulting from its isoelectric point (PI).Streptavidin is useful in affinity chromatography, ELISA, immunohistochemistry and Western Blotting.

Specifications

Conjugate HRP
Quantity 1 mg
Source Streptavidin is expressed from E.coli cells and conjugated with horseradish peroxidase under optimal conditions.
Content And Storage -20°C
Form Powder
Protein Family HRP Labeled Proteins
Buffer Na3PO4 and NaCl pH6.8
Product Type Streptavidin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.