Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 710 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Brand:  Invitrogen™ 49-4317-82
Code : Z2
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15341840
  • £161.00 / 100µg
Estimated Shipment: 23-04-2024
to see stock.
Add to basket
Alert:
Contains Azide
Specifications
0.2 mg/mL
100 μg
eBioscience™
Biotin
eFluor 710
Liquid
Streptavidin Conjugate (Fluorescent)
eFluor 710