missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 660

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Brand:  Invitrogen™ 50-4317-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381900

  • £64.20 / 25µg


EU Stock 10
Add to basket

Alert:

Contains Sodium azide


Specifications

Specifications

0.2mg/mL
Flow Cytometric Analysis, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry
2° to 8°C. Do not freeze. Light-sensitive material.
SA, SAV
eFluor 660
25μg
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Videos

For Research Use Only