Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 615 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 42-4317-80
Code : Z2
Additional Details : Weight : 0.25000kg
Alert:
Contains Sodium azide
Specifications
0.2 mg/mL | |
25 μg | |
eBioscience™ | |
Biotin |
eFluor 615 | |
Liquid | |
Streptavidin Conjugate (fluorescent) |
For Research Use Only