missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 615

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Brand:  Invitrogen™ 42-4317-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15361100

  • £184.87 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Alert:

Contains Sodium azide


Specifications

Specifications

0.2mg/mL
Immunocytochemistry, Immunohistochemical Staining of Frozen Tissue Sections, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
2° to 8°C. Do not freeze. Light-sensitive material.
SA, SAV
eFluor 615
100μg
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Videos

For Research Use Only