Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 570
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 41-4317-80
Code : NEW
Additional Details : Weight : 0.09500kg
Alert:
Contains Sodium azide
Specifications
0.2 mg/mL | |
25 μg | |
SA, Sav | |
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry |
eFluor 570 | |
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
2°C to 8°C. Do not freeze. Light-sensitive material. |
For Research Use Only