missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Straight Wall Nickel Crucible

Brand:  Alfa Aesar™ 035973.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15436145

  • £30.90 / Each


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Specifications

25mL
Outside: 45mm
Nickel Crucible
Straight Wall
22mm
Nickel
Videos
SDS