missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Straight Plug Stopper
Provide airtight covers
£317.00 - £1310.00
Product Code | Brand | Color | Material | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | For Use With (Equipment) | Price | Quantity & Availability | |||
12811144
|
DWK Life Sciences
W224100-173 |
Gray | Butyl Rubber | Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials |
£317.00
Pack of 1000 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
11788919
|
DWK Life Sciences
224100-072 |
Gray | Isoprene Rubber | Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials |
£1310.00
Pack of 1000 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||
10036671
|
DWK Life Sciences
224100-330 |
Red | Rubber | Fits 1.5cm I.D., 3cm O.D. mouth of serum bottles and vials |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Yes | |
Straight Plug Stopper |