Learn More
DWK Life Sciences Stopcock Key, threaded, SBW-J glass, ground
The most common tap plug in the ground DURAN™ single way stopcock.
Brand: DWK Life Sciences 286034208
Additional Details : Weight : 0.70000kg
Description
Solid SBW-J glass tap plugs have a thread on which the 3-part screw lock is mounted. The tap plugs are offered in 3 NS sizes and each NS size in two bore variants.
- With thread, ground
- Solid tap plug made of SBW-J glass
- The tap plugs are offered in 3 NS sizes and each NS size in two bore variants
- Assembly and thus securing of the valve plug by means of the 3-part screw locking device
- Taper 1:10
Specifications
2.5 mm | |
No | |
50/Pk. |
SBW-J Glass | |
12.5 NS |