Learn More
DWK Life Sciences Stopcock Key, threaded, SBW-J glass, ground
The most common tap plug in the ground DURAN™ single way stopcock.
Brand:  DWK Life Sciences 286037505
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 15894518
  • £367.00 / Pack of 50
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
Solid SBW-J glass tap plugs have a thread on which the 3-part screw lock is mounted. The tap plugs are offered in 3 NS sizes and each NS size in two bore variants.
  • With thread, ground
  • Solid tap plug made of SBW-J glass
  • The tap plugs are offered in 3 NS sizes and each NS size in two bore variants
  • Assembly and thus securing of the valve plug by means of the 3-part screw locking device
  • Taper 1:10
Specifications
Specifications
2.5 mm
No
50/Pk.
SBW-J Glass
14.5 NS