Learn More
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground
2-way tap plug with thread.
Brand: DWK Life Sciences 286037308
Additional Details : Weight : 3.35000kg
Description
Ground DURAN™ patent tap plugs have 2 almost parallel inclined bores.
- With thread, ground
- 2-way tap plug with 2 almost parallel inclined holes
- massive tap plug
Specifications
4 mm | |
Yes | |
25/Pk. |
Borosilicate Glass (3.3) | |
18.8 NS | |
Installation in glass apparatus and devices |