Learn More
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground
2-way tap plug with thread.
Brand:  DWK Life Sciences 286036706
Additional Details : Weight : 12.50000kg
 View more versions of this product
Product Code. 15864518
  • £865.00 / Pack of 50
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
Ground DURAN™ patent tap plugs have 2 almost parallel inclined bores.
  • With thread, ground
  • 2-way tap plug with 2 almost parallel inclined holes
  • massive tap plug
Specifications
Specifications
2.5 mm
Yes
50/Pk.
Borosilicate Glass (3.3)
14.5 NS
Installation in glass apparatus and devices