Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
For use with ground DURAN™ single way stopcocks.
Brand:  DWK Life Sciences 286036209
Additional Details : Weight : 6.25000kg
 View more versions of this product
Product Code. 15844518
  • £439.00 / Pack of 25
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
  • With groove, polished
  • The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
  • Solid glass tap plugs
  • Taper 1:10
Specifications
Specifications
8.0 mm
Yes
25/Pk.
Borosilicate Glass (3.3)
24.0 NS