Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
For use with ground DURAN™ single way stopcocks.
Brand: DWK Life Sciences 286036106
Additional Details : Weight : 12.50000kg
Description
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
- With groove, polished
- The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
- Solid glass tap plugs
- Taper 1:10
Specifications
6.0 mm | |
Yes | |
25/Pk. |
Borosilicate Glass (3.3) | |
21.5 NS |