Learn More
DWK Life Sciences DURAN™ Burette Stopcock Key, side entry, drilled and ground, with three part retaining device
For use in burettes.
Brand:  DWK Life Sciences 286035504
Additional Details : Weight : 5.00000kg
Product Code. 15824518
  • £590.00 / Pack of 50
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
Lateral DURAN™ burette tap Plug with 3-piece tap locking have a calibrated tip (Øi 0.80-0.85 mm).
  • For the lateral burette outlet e.g. for automatic burettes
  • Burette stopcock key with calibrated tip (Øi 0,80-0,85 mm) for accurate working
  • Ground tap plug, with 3-part tap safety catch
Specifications
Specifications
2.5 mm
Yes
50/Pk.
Borosilicate Glass (3.3), Polypropylene
12.5 NS
Installation in glass apparatus and devices