Learn More
DWK Life Sciences DURAN™ Glass Stopcock for Aspirator Bottles, Standard Taper Ground Cone
Stopcock with glass key and a ground cone for Aspirator bottles
Brand: DWK Life Sciences 241480401
Code : A0
Additional Details : Weight : 0.09100kg
Description
Aspirator stopcock with glass key and a ground cone to fit the socket side-arm of DURAN™ aspirator bottles.
- All glass construction
- Tight closure due to standard ground joint
Specifications
5000 to 20,000 mL | |
Controlled dispensing of liquids and reagents | |
Not Autoclavable |
Borosilicate Glass (3.3) | |
1/Pk. |