missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK38L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558471
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: STVGTPDYIA PEVFMQTGYN KLCDWWSLGV IMYEMLIGYP PFCSETPQET YRKVMNWKET LVFPPEVPIS EKAKDL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
STK38L Polyclonal antibody specifically detects STK38L in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
STK38L | |
Polyclonal | |
Unconjugated | |
Stk38l | |
4930473A22Rik; B230328I19; KIAA0965; Ndr2; NDR2 protein kinase; Ndr54; nuclear Dbf2-related 2; nuclear Dbf2-related kinase 2; RGD1564793; serine/threonine kinase 38 like; serine/threonine kinase 38-like; serine/threonine-protein kinase 38-like; STK38L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2H1 | |
Stk38l | |
Recombinant protein corresponding to Human STK38L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only