missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550835
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total STK32A protein.
STK32A Polyclonal antibody specifically detects STK32A in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
STK32A | |
Polyclonal | |
Unconjugated | |
STK32A | |
A930015B13Rik; CTB-108O6.2; serine/threonine kinase 32A; serine/threonine-protein kinase 32A; Stk32a; YANK1; Yet another novel kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202374 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8WU08 | |
STK32A | |
Synthetic peptide corresponding to residues near the C terminal of human serine/threonine kinase 32A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |