missing translation for 'onlineSavingsMsg'
Learn More

STK17A (DRAK1) Recombinant Human Protein

STK17A (DRAK1) Recombinant Human Protein

Brand:  Invitrogen PV3783

Code : 50

Additional Details : Weight : 0.05250kg

Product Code. 11553527

  • £434.00 / 10µg
Estimated Shipment: 01-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

STK17A (DRAK1) is a serine⁄threonine kinase localized in the nucleus and involved in apoptosis.
Specifications

Specifications

NP_004751
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
74kDa
10μg
For research use only. Not for use in diagnostic procedures
STK17A (DRAK1)
STK17A (DRAK1) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 290μg/mL MLCK peptide substrate (KKRAARATSDVFA-NH2) and trace [32P]-γ-ATP for 10 minutes at 30°C
STK17A (DRAK1) Recombinant Human Protein
Human
Baculovirus
Serine/threonine kinase
0.25mg/mL
554, 9263
STK17A (DRAK1) Recombinant Human Protein
–80°C
1 Each
STK17A (DRAK1) Recombinant Human Protein
DAPK
36nmoles of phosphate transferred to MLCK peptide substrate (KKRAARATSDVFA-NH2) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
GST Tag
Kinase and inhibitors
75% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.