missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Magnetic Stirring Bar with Pivot Ring

Create high turbulence even at a low speed with a stirring bar that features and angled surface. Kartell™ Magnetic Stirring Bar with Pivot Ring reduces the contact surface and fosters optimal stir bar positioning.

Brand:  Kartell™ 799

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10498832

  • £84.30 / Pack of 10


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

White
Pivot ring
Stirring bar
70.78mm
Magnet PTFE Coated
10/Pk.
9.22mm
PTFE (Polytetrafluoroethylene)
Videos