missing translation for 'onlineSavingsMsg'
Learn More

STF 31, Tocris Bioscience™

NAMPT inhibitor; also GLUT1 inhibitor

Brand:  RD Systems (Bio-Techne) 4484/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464824

  • £848.19 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of GLUT1; inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells. Activity causes necrotic cell death in von Hippel-Lindau (VHL)-deficient RCC cells. Also NAMPT inhibitor. Eliminates human pluripotent stem cells from culture with limited toxicity towards differentiated cells.
Specifications

Chemical Identifiers

724741-75-7
423.531
stf-31, stf 31, 4-4-1,1-dimethylethyl phenyl sulfonyl amino methyl-n-3-pyridinylbenzamide, stf31, stf hplc, 4-4-tert-butylphenylsulfonamido methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylphenyl sulfonyl amino methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylbenzenesulfonamido methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylphenyl sulfonylamino methyl-n-pyridin-3-ylbenzamide, 2hj
4-[[(4-tert-butylphenyl)sulfonylamino]methyl]-N-pyridin-3-ylbenzamide
C23H25N3O3S
NGQPRVWTFNBUHA-UHFFFAOYSA-N
984333
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NCC2=CC=C(C=C2)C(=O)NC3=CN=CC=C3

Specifications

50mg
724741-75-7
stf-31, stf 31, 4-4-1,1-dimethylethyl phenyl sulfonyl amino methyl-n-3-pyridinylbenzamide, stf31, stf hplc, 4-4-tert-butylphenylsulfonamido methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylphenyl sulfonyl amino methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylbenzenesulfonamido methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylphenyl sulfonylamino methyl-n-pyridin-3-ylbenzamide, 2hj
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NCC2=CC=C(C=C2)C(=O)NC3=CN=CC=C3
423.531
423.53
STF 31
C23H25N3O3S
NGQPRVWTFNBUHA-UHFFFAOYSA-N
4-[[(4-tert-butylphenyl)sulfonylamino]methyl]-N-pyridin-3-ylbenzamide
984333
>99%
Videos
Documents