missing translation for 'onlineSavingsMsg'
Learn More

Merck Steritop™ Threaded Bottle Top Filters

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.

Brand:  Merck S2GPT02RE

109.50 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Additional Details : Weight : 1.73700kg

 View more versions of this product

Product Code. 15929180

  • £126.00 / Pack of 12
In Stock
to see stock.
Add to basket

Description

Description

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.

Order Info

Note: Image above shows Bottle Top Filter with receiver flask (sold separately).

Specifications

Specifications

Clear with blue adapter
Polystyrene
0.22 μm
Sterile filtration
Bottle Top Filter
95 mm
45 mm
Single-use
PES
Sterile
45 mm
250 mL
95 mm
12.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers