missing translation for 'onlineSavingsMsg'
Learn More

0 Sterilization Bags with Plain Closure

Brand:  0 V1PC

UNSPSC : 42281916

Code : 03

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12719105

  • £116.00 / Pack of 1000


Estimated Shipment
05-07-2017
Add to basket

Specifications Description & Specifications

Specifications

CE Marked
Suitable For Steam and ETO Sterilisation, amd formaldehyde
1000/pk
30mm
Plain
150mm
125mm